missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100P Protein

A cDNA sequence encoding the S100P was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13408-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15903479

  • 5195.00€ / 1mg

Spedizione stimata: 03-04-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

S100P Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Phosphate buffer pH 7.2, 0.1M NaCl.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MTELEAAMGM IIDVFSRYSG SEGSTQTLTK GELKVLMEKE LPGFLQSGKD KDAVDKLLKD LDANGDAQVD FSEFIVFVAA ITSACHKYFE KAGLK
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali