missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SDHAF2 Protein

A cDNA sequence encoding the SDHAF2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13442-20ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943579

Quantità Prezzo
1 166.00€ / 20µg
10 155.97€ / 20µg
Spedizione stimata
11-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

SDHAF2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SDHAF2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 30% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSFRRFYR GDSPTDSQKD MIEIPLPPWQ ERTDESIETK RARLLYESRK RGMLENCILL SLFAKEHLQH MTEKQLNLYD RLINEPSNDW DIYYWATEAK PAPEIFENEV MALLRDFAKN KNKEQRLRAP DLEYLFEKPR
Videos
SDS
Documenti