missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine--pyruvate aminotransferase Protein

A DNA sequence encoding the Homo sapiens (Human) Serine--pyruvate aminotransferase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5627-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15956539

  • 458.00€ / 100µg

Spedizione stimata: 27-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

NP_000021.1
Serine--pyruvate aminotransferase Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
189
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Agxt
Recombinant Protein
E. coli
MASHKLLVTP PKALLKPLSI PNQLLLGPGP SNLPPRIMAA GGLQMIGSMS KDMYQIMDEI KEGIQYVFQT RNPLTLVISG SGHCALEAAL VNVLEPGDSF LVGANGIWGQ RAVDIGERIG ARVHPMTKDP GGHYTLQEVE EGLAQHKPVL LFLTHGESST GVLQPLDGFG ELCHRYKCLL LVDSVASLGG TPLYMDRQGI DILYSGSQKA LNAPPGTSLI SFSDKAKKKM YSRKTKPFSF YLDIKWLANF WGCDDQPRMY HHTIPVISLY SLRESLALIA EQGLENSWRQ HREAAAYLHG RLQALGLQLF VKDPALRLPT VTTVAVPAGY DWRDIVSYVI DHFDIEIMGG LGPSTGKVLR IGLLGCNATR ENVDRVTEAL RAALQHCPKK KL
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali