missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sialidase-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Sialidase-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8892-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_006647.3 | |
Sialidase-3 Protein | |
Research Use Only | |
NEU3 | |
Recombinant Protein | |
E. coli | |
EEVTTCSFNS PLFRQEDDRG ITYRIPALLY IPPTHTFLAF AEKRSTRRDE DALHLVLRRG LRIGQLVQWG PLKPLMEATL PGHRTMNPCP VWEQKSGCVF LFFICVRGHV TERQQIVSGR NAARLCFIYS QDAGCSWSEV RDLTEEVIGS ELKHWATFAV GPGHGIQLQS GRLVIPAYTY YIPSWFFCFQ LPCKTRPHSL MIYSDDLGVT WHHGRLIRPM VTVECEVAEV TGRAGHPVLY CSARTPNRCR AEALSTDHGE GFQRLALSRQ LCEPPHGCQG SVVSFRPLEI PHRCQDSSSK DAPTIQQSSP GSSLRLEEEA GTPSESWLLY SHPTSRKQRV DLGIYLNQTP LEAACWSRPW ILHCGPCGYS DLAALEEEGL FGCLFECGTK QECEQIAFRL FTHREILSHL QGDCTSPGRN PSQFKSN | |
Greater than 90% as determined by SDS-PAGE. |
10825 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |