missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Src kinase-associated phosphoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Src kinase-associated phosphoprotein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7785-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001068567.1 | |
Src kinase-associated phosphoprotein 1 | |
Research Use Only | |
SKAP1 | |
Recombinant Protein | |
E. coli | |
MQAAALPEEI RWLLEDAEEF LAEGLRNENL SAVARDHRDH ILRGFQQIKA RYYWDFQPQG GDIGQDSSDD NHSGTLGLSL TSDAPFLSDY QDEGMEDIVK GAQELDNVIK QGYLEKKSKD HSFFGSEWQK RWCVVSRGLF YYYANEKSKQ PKGTFLIKGY GVRMAPHLRR DSKKESCFEL TSQDRRSYEF TATSPAEARD WVDQISFLLK DLSSLTIPYE EDEEEEEKEE TYDDIDGFDS PSCGSQCRPT ILPGSVGIKE PTEEKEEEDI YEVLPDEEHD LEEDESGTRR KGVDYASYYQ GLWDCHGDQP DELSFQRGDL IRILSKEYNM YGWWVGELNS LVGIVPKEYL TTAFEVEE | |
Greater than 90% as determined by SDS-PAGE. |
8631 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |