missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Stathmin-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Stathmin-4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8172-ec-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16063051

Quantità Prezzo
1 675.00€ / 200µg
5 629.51€ / 200µg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001269982.1
Stathmin-4 Protein
200 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
81551
Greater than 80% as determined by SDS-PAGE.
Research Use Only
STMN4
Recombinant Protein
E. coli
MTLAAYKEKM KELPLVSLFC SCFLADPLNK SSYKYEGWCG RQCRRKDESQ RKDSADWRER RAQADTVDLN WCVISDMEVI ELNKCTSGQS FEVILKPPSF DGVPEFNASL PRRRDPSLEE IQKKLEAAEE RRKYQEAELL KHLAEKREHE REVIQKAIEE NNNFIKMAKE KLAQKMESNK ENREAHLAAM LERLQEKDKH AEEVRKNKEL KEEASR
Videos
Documenti