missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCL1A Protein
A cDNA sequence encoding the TCL1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13711-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TCL1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAECPTLGEA VTDHPDRLWA WEKFVYLDEK QHAWLPLTIE IKDRLQLRVL LRREDVVLGR PMTPTQIGPS LLPIMWQLYP DGRYRSSDSS FWRLVYHIKI DGVEDMLLEL LPDD | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TCL1A protein solution contains 50mM Tris-HCl buffer (pH 7.5) and 10% glycerol. |