missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin-interacting Protein
A DNA sequence encoding the Homo sapiens(Human) Thioredoxin-interacting protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10023-iv-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001300901.1 | |
Thioredoxin-interacting Protein | |
Research Use Only | |
Txnip | |
Recombinant Protein | |
E. coli | |
MVMFKKIKSF EVVFNDPEKV YGSGEKVAGR VIVEVCEVTR VKAVRILACG VAKVLWMQGS QQCKQTSEYL RYEDTLLLED QPTGENEMVI MRPGNKYEYK FGFELPQGPL GTSFKGKYGC VDYWVKAFLD RPSQPTQETK KNFEVVDLVD VNTPDLMAPV SAKKEKKVSC MFIPDGRVSV SARIDRKGFC EGDEISIHAD FENTCSRIVV PKAAIVARHT YLANGQTKVL TQKLSSVRGN HIISGTCASW RGKSLRVQKI RPSILGCNIL RVEYSLLIYV SVPGSKKVIL DLPLVIGSRS GLSSRTSSMA SRTSSEMSWV DLNIPDTPEA PPCYMDVIPE DHRLESPTTP LLDDMDGSQD SPIFMYAPEF KFMPPPTYTE VDPCILNNNV Q | |
Greater than 85% as determined by SDS-PAGE. |
10628 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |