missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNF-alpha Protein
A DNA sequence encoding the Human TNF-alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10352-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000585.2 | |
TNF-alpha Protein | |
Research Use Only | |
TNF | |
The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg in the presence of actinomycin D. | |
Human | |
His | |
0.2 m filtered PBS, pH 7.0, lyophilized |
7124 | |
50 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MHHHHHHVRS SSRTPSDKPV AHVVANPQAE GQLQWLNRRA NALLANGVEL RDNQLVVPSE GLYLIYSQVL FKGQGCPSTH VLLTHTISRI AVSYQTKVNL LSAIKSPCQR ETPEGAEAKP WYEPIYLGGV FQLEKGDRLS AEINRPDYLD FAESGQVYFG IIAL | |
Greater than 97% as determined by SDS-PAGE and HPLC. |