missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transmembrane protease serine 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Transmembrane protease serine 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9439-ye-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16058311

Quantità Prezzo
1 2645.00€ / 1mg
3 2469.90€ / 1mg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001128571.1
Transmembrane protease serine 2 Protein
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
7113
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TMPRSS2
Recombinant Protein
Yeast
WKFMGSKCSN SGIECDSSGT CINPSNWCDG VSHCPGGEDE NRCVRLYGPN FILQVYSSQR KSWHPVCQDD WNENYGRAAC RDMGYKNNFY SSQGIVDDSG STSFMKLNTS AGNVDIYKKL YHSDACSSKA VVSLRCIACG VNLNSSRQSR IVGGESALPG AWPWQVSLHV QNVHVCGGSI ITPEWIVTAA HCVEKPLNNP WHWTAFAGIL RQSFMFYGAG YQVEKVISHP NYDSKTKNND IALMKLQKPL TFNDLVKPVC LPNPGMMLQP EQLCWISGWG ATEEKGKTSE VLNAAKVLLI ETQRCNSRYV YDNLITPAMI CAGFLQGNVD SCQGDSGGPL VTSKNNIWWL IGDTSWGSGC AKAYRPGVYG NVMVFTDWIY RQMRADG
Videos
Documenti