missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Trypsin-3 / PRSS3 Protein

A DNA sequence encoding the Homo sapiens (Human) Trypsin-3 / PRSS3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8742-ec-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16056251

Quantità Prezzo
1 675.00€ / 200µg
5 629.51€ / 200µg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001184026.2
Trypsin-3 / PRSS3 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
5646
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PRSS3
Recombinant Protein
E. coli
IVGGYTCEEN SLPYQVSLNS GSHFCGGSLI SEQWVVSAAH CYKTRIQVRL GEHNIKVLEG NEQFINAAKI IRHPKYNRDT LDNDIMLIKL SSPAVINARV STISLPTTPP AAGTECLISG WGNTLSFGAD YPDELKCLDA PVLTQAECKA SYPGKITNSM FCVGFLEGGK DSCQRDSGGP VVCNGQLQGV VSWGHGCAWK NRPGVYTKVY NYVDWIKDTI AAN
Videos
Documenti