missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Unconventional myosin-XIX Protein

A DNA sequence encoding the Homo sapiens (Human) Unconventional myosin-XIX, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7873-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16061301

  • 2385.00€ / 1mg
Spedizione stimata: 29-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

NP_001028752.1
Unconventional myosin-XIX Protein
Research Use Only
MYO19
Recombinant Protein
E. coli
LQQVNGHNPG SDGQAREYLR EDLQEFLGGE VLLYKLDDLT RVNPVTLETV LRCLQARYMA DTFYTNAGCT LVALNPFKPV PQLYSPELMR EYHAAPQPQK LKPHVFTVGE QTYRNVKSLI EPVNQSIVVS GESGAGKTWT SRCLMKFYAV VATSPASWES HKIAERIEQR ILNSNPVMEA FGNACTLRNN NSSRFGKFIQ LQLNRAQQMT GAAVQTYLLE KTRVACQASS ERNFHIFYQI CKGASEDERL QWHLPEGAAF SWLPNPERSL EEDCFEVTRE AMLHLGIDTP TQNNIFKVLA GLLHLGNIQF AASEDEAQPC QPMDDAKYSV RTAASLLGLP EDVLLEMVQI RTIRAGRQQQ VFRKPCARAE CDTRRDCLAK LIYARLFDWL VSVINSSICA DTDSWTTFIG LLDVYGFESF PDNSLEQLCI NYANEKLQLE ACGLVETIHI SAAGFPIRVS HRNFVERYKL LRRLHPCTSS GPDSPYPAKG LPEWCPHSEE ATLEPLIQDI LHTLPVLTQA AAITGDSAEA MPAPMHCGRT KVFMTDSMLE LLECGRARVL EQCARCIQGG WRRHRHREQE RQWRAVMLIQ AAIRSWLTRK HIQRLHAAAT VIKRAWQKWR IRMACLAAKE LDGVEEKHFS QAPCSLSTSP LQTRLLEAII RLWPLGLVLA NTAMGVGSFQ RKLVVWACLQ LPRGSPSSYT VQTAQDQAGV TSIRALPQGS IKFHCRKSPL RYADICPEPS PYSITGFNQI LLERHRLIHV TSSAFTGLG
Greater than 90% as determined by SDS-PAGE.
80179
1 mg
Not determined.
Full Length of Isoform 4
Human
His-SUMO
Tris-based buffer, 50% glycerol
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali