missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAMP3 Protein
A cDNA sequence encoding the VAMP3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13917-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
VAMP3 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The VAMP3 protein solution contains 20mM Tris pH 7.5 and 10% glycerol. |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSTGPTAATG SNRRLQQTQN QVDEVVDIMR VNVDKVLERD QKLSELDDRA DALQAGASQF ETSAAKLKRK YWWKNCK |