missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vesicle-associated membrane protein 1
A DNA sequence encoding the Homo sapiens (Human) Vesicle-associated membrane protein 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6876-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001138621.1 | |
Vesicle-associated membrane protein 1 | |
Research Use Only | |
VAMP7 | |
Recombinant Protein | |
E. coli | |
AILFAVVARG TTILAKHAWC GGNFLEVTEQ ILAKIPSENN KLTYSHGNYL FHYICQDRIV YLCITDDDFE RSRAFNFLNE IKKRFQTTYG SRAQTALPYA MNSEFSSVLA AQLKHHSENK GLDKVMETQA QVDELKGIMV RNIDLVAQRG ERLELLIDKT ENLVDSSVTF KTTSRNLARA MCMKN | |
Greater than 90% as determined by SDS-PAGE. |
6845 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |