missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 638 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 638, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6910-ec-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16056700

Quantità Prezzo
1 1180.00€ / 500µg
3 1100.47€ / 500µg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001014972.1
Zinc finger protein 638 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
27332
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ZNF638
Recombinant Protein
E. coli
TFATLNTKGN EGDTVRDSIG FISSQVPEDP STLVTVDEIQ DDSSDLHLVT LDEVTEEDED SLADFNNLKE ELNFVTVDEV GEEEDGDNDL KVELAQSKND HPTDKKGNRK KRAVDTKKTK LESLSQVGPV NENVMEEDLK TMIERHLTAK TPTKRVRIGK TLPSEKAVVT EPAKGEEAFQ MSEVDEESGL KDSEPERKRK KTEDSSSGKS VASDVPEELD FLVPKAGFFC PICSLFYSGE KAMTNHCKST RHKQNTEKFM AKQRKEKEQN EAEERSSR
Videos
Documenti