missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZMAT3 Protein
A cDNA sequence encoding the ZMAT3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP14002-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
64393 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
ZMAT3 | |
Human | |
His | |
The ZMAT3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 50% glycerol. |
ZMAT3 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMILLQHA VLPPPKQPSP SPPMSVATRS TGTLQLPPQK PFGQEASLPL AGEEELSKGG EQDCALEELC KPLYCKLCNV TLNSAQQAQA HYQGKNHGKK LRNYYAANSC PPPARMSNVV EPAATPVVPV PPQMGSFKPG GRVILATEND YCKLCDASFS SPAVAQAHYQ GKNHAKRLRL AEAQSNSFSE SSELGQRRAR KEGNEFKMMP NRRNMYTVQN NSAGPYFNPR SRQRIPRDLA MCVTPSGQFY CSMCNVGAGE EMEFRQHLES KQHKSKVSEQ RYRNEMENLG YV |