missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza B (strain B / Lee / 1940) NucleoProtein
A DNA sequence encoding the Influenza B virus (strain B/Lee/1940) Influenza B (strain B/Lee/1940) Nucleoprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7357-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_056661.1 | |
Influenza B (strain B / Lee / 1940) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
Yeast | |
MSNMDIDSIN TGTIDKTPEE LTPGTSGATR PIIKPATLAP PSNKRTRNPS PERTTTSSET DIGRKIQKKQ TPTEIKKSVY KMVVKLGEFY NQMMVKAGLN DDMERNLIQN AQAVERILLA ATDDKKTEYQ KKRNARDVKE GKEEIDHNKT GGTFYKMVRD DKTIYFSPIK ITFLKEEVKT MYKTTMGSDG FSGLNHIMIG HSQMNDVCFQ RSKGLKRVGL DPSLISTFAG STLPRRSGTT GVAIKGGGTL VDEAIRFIGR AMADRGLLRD IKAKTAYEKI LLNLKNKCSA PQQKALVDQV IGSRNPGIAD IEDLTLLARS MVVVRPSVAS KVVLPISIYA KIPQLGFNTE EYSMVGYEAM ALYNMATPVS ILRMGDDAKD KSQLFFMSCF GAAYEDLRVL SALTGTEFKP RSALKCKGFH VPAKEQVEGM GAALMSIKLQ FWAPMTRSGG NEVSGEGGSG QISCSPVFAV ERPIALSKQA VRRMLSMNVE GRDADVKGNL LKMMNDSMAK KTSGNAFIGK KMFQISDKNK VNPIEIPIKQ TIPNFFFGRD TAEDYDDLDY | |
Greater than 90% as determined by SDS-PAGE. |
26824002 | |
200 μg | |
Not determined. | |
Full Length | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |