missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis MPT51 / MPB51 antigen Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) MPT51/MPB51 antigen, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7383-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
WP_003420783.1 | |
200 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
M. tuberculosis MPT51 / MPB51 antigen Protein | |
Research Use Only | |
mpt51 | |
Recombinant Protein | |
E. coli | |
AEPTAKAAPY ENLMVPSPSM GRDIPVAFLA GGPHAVYLLD AFNAGPDVSN WVTAGNAMNT LAGKGISVVA PAGGAYSMYT NWEQDGSKQW DTFLSAELPD WLAANRGLAP GGHAAVGAAQ GGYGAMALAA FHPDRFGFAG SMSGFLYPSN TTTNGAIAAG MQQFGGVDTN GMWGAPQLGR WKWHDPWVHA SLLAQNNTRV WVWSPTNPGA SDPAAMIGQA AEAMGNSRMF YNQYRSVGGH NGHFDFPASG DNGWGSWAPQ LGAMSGDIVG AIR | |
Greater than 90% as determined by SDS-PAGE. |