missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Steroid C26-monooxygenase Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Steroid C26-monooxygenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7253-ye-1mg

Additional Details : Peso : 0.01000kg

 View more versions of this product

Product Code. 16057870

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Description

Description

Specifications

Specifications

NP_218062.1
M. tuberculosis Steroid C26-monooxygenase Protein
Research Use Only
cyp125
Recombinant Protein
Yeast
MSWNHQSVEI AVRRTTVPSP NLPPGFDFTD PAIYAERLPV AEFAELRSAA PIWWNGQDPG KGGGFHDGGF WAITKLNDVK EISRHSDVFS SYENGVIPRF KNDIAREDIE VQRFVMLNMD APHHTRLRKI ISRGFTPRAV GRLHDELQER AQKIAAEAAA AGSGDFVEQV SCELPLQAIA GLLGVPQEDR GKLFHWSNEM TGNEDPEYAH IDPKASSAEL IGYAMKMAEE KAKNPADDIV TQLIQADIDG EKLSDDEFGF FVVMLAVAGN ETTRNSITQG MMAFAEHPDQ WELYKKVRPE TAADEIVRWA TPVTAFQRTA LRDYELSGVQ IKKGQRVVMF YRSANFDEEV FQDPFTFNIL RNPNPHVGFG GTGAHYCIGA NLARMTINLI FNAVADHMPD LKPISAPERL RSGWLNGIKH WQVDYTGRCP VAH
Greater than 90% as determined by SDS-PAGE.
887782
1 mg
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers