missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque FLT3L / Flt3 ligand Protein

A DNA sequence encoding the Rhesus Macaque FLT3L / Flt3 ligand, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10447-ec-2ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943888

Quantità Prezzo
1 105.00€ / 2µg
10 98.00€ / 2µg
Spedizione stimata
06-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

XP_014980066.1
FLT3L / Flt3 ligand Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVPSNLQD EELCGALWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQHP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPRSPGALEA TALTAPQRP
719239
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
FLT3LG
The ED50 as determined by a cell proliferation assay using human AML5 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Macaque
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documenti