missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bovine Enterokinase / PRSS7 Protein
A DNA sequence encoding the Mus musculus (Mouse) Bovine Enterokinase / PRSS7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6539-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_032967.1 | |
Enterokinase / PRSS7 Protein | |
Research Use Only | |
Tmprss15 | |
Recombinant Protein | |
E. coli | |
IVGGSDAQAG AWPWVVALYH RDRSTDRLLC GASLVSSDWL VSAAHCVYRR NLDPTRWTAV LGLHMQSNLT SPQVVRRVVD QIVINPHYDR RRKVNDIAMM HLEFKVNYTD YIQPICLPEE NQIFIPGRTC SIAGWGYDKI NAGSTVDVLK EADVPLISNE KCQQQLPEYN ITESMICAGY EEGGIDSCQG DSGGPLMCQE NNRWFLVGVT SFGVQCALPN HPGVYVRVSQ FIEWIHSFLH | |
Greater than 80.8% as determined by SDS-PAGE. |
19146 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |