missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Calcium-dependent phospholipase A2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6499-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001116426.1 | |
Calcium-dependent phospholipase A2 Protein | |
Research Use Only | |
PLA2G5 | |
Recombinant Protein | |
E. coli | |
GLLELKSMIE KVTGKNAFKN YGFYGCYCGW GGRGTPKDGT DWCCQMHDRC YGQLEEKDCA IRTQSYDYRY TNGLVICEHD SFCPMRLCAC DRKLVYCLRR NLWTYNPLYQ YYPNFLC | |
Greater than 90% as determined by SDS-PAGE. |
18784 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |