missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CST3 Protein
A cDNA sequence encoding the CST3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11541-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
13010 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASATPKQG PRMLGAPEEA DANEEGVRRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQL VAGVNYFLDV EMGRTTCTKS QTNLTDCPFH DQPHLMRKAL CSFQIYSVPW KGTHSLTKFSCKNA | |
Greater than 95% as determined by SDS-PAGE. |
CST3 Protein | |
Research Use Only | |
Cst3 | |
Mouse | |
His | |
The sterile filtered concentrated (0.5 mg/ml) protein solution was lyophilized with 20mM Tris and 50mM NaCl pH 7.5. |