missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL1 / MGSA / NAP-3 Protein
A DNA sequence encoding the Mouse CXCL1 / MGSA / NAP-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10248-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_032202.1 | |
CXCL1 / MGSA / NAP-3 Protein | |
Research Use Only | |
CXCL1 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood neutrophils is in a concentration range of 10-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
14825 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APIANELRCQ CLQTMAGIHL KNIQSLKVLP SGPHCTQTEV IATLKNGREA CLDPEAPLVQ KIVQKMLKGV PK | |
Greater than 97% as determined by SDS-PAGE and HPLC. |