Learn More
enQuireBio™ Recombinant Mouse Frataxin, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Frataxin, mitochondrial, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP6059-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15939689
Quantità Prezzo
1 245.00€ / 10µg
6 228.51€ / 10µg
Spedizione stimata
18-01-2021
Specifica
NP_032070.1
Frataxin, mitochondrial Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
14297
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Fxn
Recombinant Protein
Yeast
LGTLDNPSSL DETAYERLAE ETLDSLAEFF EDLADKPYTL EDYDVSFGDG VLTIKLGGDL GTYVINKQTP NKQIWLSSPS SGPKRYDWTG KNWVYSHDGV SLHELLAREL TKALNTKLDL SSLAYSGKGT