missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IDH1 Protein
A DNA sequence encoding the Mus musculus (Mouse) IDH1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6192-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001104790.1 | |
IDH1 Protein | |
Research Use Only | |
Idh1 | |
Recombinant Protein | |
E. coli | |
GGSVVEMQGD EMTRIIWELI KEKLILPYVE LDLHSYDLGI ENRDATNDQV TKDAAEAIKK YNVGVKCATI TPDEKRVEEF KLKQMWKSPN GTIRNILGGT VFREAIICKN IPRLVTGWVK PIIIGRHAYG DQYRATDFVV PGPGKVEITY TPKDGTQKVT YMVHDFEEGG GVAMGMYNQD KSIEDFAHSS FQMALSKGWP LYLSTKNTIL KKYDGRFKDI FQEIYDKKYK SQFEAQKICY EHRLIDDMVA QAMKSEGGFI WACKNYDGDV QSDSVAQGYG SLGMMTSVLI CPDGKTVEAE AAHGTVTRHY RMYQKGQETS TNPIASIFAW SRGLAHRAKL DNNTELSFFA KALEDVCIET IEAGFMTKDL AACIKGLPNV QRSDYLNTFE FMDKLGENLK A | |
Greater than 90% as determined by SDS-PAGE. |
15926 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |