missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-17F (Mammalian-expressed) Protein

Codice prodotto. 18778596
Cambia vista
Click to view available options
Quantità:
50 μg
Dimensione della confezione:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18778596 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18778596 Fornitore R&D Systems™ N. del fornitore 10424IL050
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells.

The ED50 for this effect is 40-240 ng/mL.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso Q7TNI7.1
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS.
ID gene (immissione) 112744
Peso molecolare Observed: 17-27 kDa, under reducing conditions; Theroretical: 15 kDa (monomer)
Quantità 50 μg
Sorgente Mouse myeloma cell line, NS0-derived mouse IL-17 F protein Arg29 - Ala161
Requisiti di stoccaggio Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Concentrazione di endotossine <0.10 EU / 1 μg of the protein by the LAL method.
Coniugato Unconjugated
Ricostituzione Reconstitute at 200 μg/mL in PBS.
Linea di prodotti R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Proteine IL-17 F
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.