missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LAIR1 Protein
A cDNA sequence encoding the LAIR1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12525-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
LAIR1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPQEGSLPD ITIFPNSSLM ISQGTFVTVV CSYSDKHDLY NMVRLEKDGS TFMEKSTEPY KTEDEFEIGP VNETITGHYS CIYSKGITWS ERSKTLELKV IKENVIQTPA PGPTSDTSWL KTYSIYHHHH HH | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
LAIR1 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |