missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Osteocalcin Protein
A DNA sequence encoding the Mus musculus (Mouse) Osteocalcin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5716-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_031567.1 | |
Osteocalcin Protein | |
Research Use Only | |
Bglap | |
Recombinant Protein | |
E. coli | |
YLGASVPSPD PLEPTREQCE LNPACDELSD QYGLKTAYKR IYGITI | |
Greater than 90% as determined by SDS-PAGE. |
12096 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |