missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Pituitary homeobox 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Pituitary homeobox 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6496-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_032878.1 | |
Pituitary homeobox 3 Protein | |
Research Use Only | |
Pitx3 | |
Recombinant Protein | |
Yeast | |
MEFGLLGEAE ARSPALSLSD AGTPHPPLPE HGCKGQEHSD SEKASASLPG GSPEDGSLKK KQRRQRTHFT SQQLQELEAT FQRNRYPDMS TREEIAVWTN LTEARVRVWF KNRRAKWRKR ERSQQAELCK GGFAAPLGGL VPPYEEVYPG YSYGNWPPKA LAPPLAAKTF PFAFNSVNVG PLASQPVFSP PSSIAASMVP SAAAAPGTVP GPGALQGLGG APPGLAPAAV SSGAVSCPYA SAAAAAAAAA SSPYVYRDPC NSSLASLRLK AKQHASFSYP AVPGPPPAAN LSPCQYAVER PV | |
Greater than 90% as determined by SDS-PAGE. |
18742 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |