missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein flightless-1 homolog Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein flightless-1 homolog, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9918-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Spezifikation
NP_071292.1 | |
Protein flightless-1 homolog Protein | |
Research Use Only | |
Flii | |
Recombinant Protein | |
Yeast | |
VGQLPGLTIW QIENFVPVLV EEAFHGKFYE ADCYIVLKTF LDDSGSLNWE IYYWIGGEAT LDKKACSAIH AVNLRNYLGA ECRTVREEMG DESEEFLQVF DNDISYIEGG TASGFYTVED THYVTRMYRV YGKKNIKLEP VPLKGSSLDP RFVFLLDQGL DIYVWRGAQA TLSNTTKARL FAEKINKNER KGKAEITLLV QGQEPPGFWD VLGGEPSEIK NHVPDDFWPP QPKLYKVGLG LGYLELPQIN YKLSVEHKKR PKVELMPGMR LLQSLLDTRC VYILDCWSDV FIWLGRKSPR LVRAAALKLG QELCGMLHRP RHTVVSRSLE GTE | |
Greater than 90% as determined by SDS-PAGE. |
14248 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |