missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A6 Protein
A cDNA sequence encoding the S100A6 was constructed and used to recombinantly synthesize the protein.
181.00€ - 3595.00€
Specifica
ID gene (immissione) | 20200 |
---|---|
Nome | S100A6 Protein |
Purezza | Greater than 90% as determined by SDS-PAGE. |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15943439
|
enQuireBio™
QP13397-HIS-20UG |
20 μg |
181.00€
20µg |
Eseguire il login per visualizzare lo stock disponibile |
15933439
|
enQuireBio™
QP13397-HIS-1MG |
1 mg |
3595.00€
1mg |
Eseguire il login per visualizzare lo stock disponibile |
Specifica
20200 | |
Greater than 90% as determined by SDS-PAGE. | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MACPLDQAIG LLVAIFHKYS GKEGDKHTLS KKELKELIQK ELTIGSKLQD AEIARLMDDL DRNKDQEVNF QEYVAFLGAL ALIYNEALK |
S100A6 Protein | |
Research Use Only | |
S100a6 | |
Mouse | |
His | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing no additives. |