missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum amyloid A-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-3 protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7359-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_035445.1 | |
Serum amyloid A-3 Protein | |
Research Use Only | |
Saa3 | |
Recombinant Protein | |
Yeast | |
RWVQFMKEAG QGSRDMWRAY SDMKKANWKN SDKYFHARGN YDAARRGPGG AWAAKVISDA REAVQKFTGH GAEDSRADQF ANEWGRSGKD PNHFRPAGLP KRY | |
Greater than 80% as determined by SDS-PAGE. |
20210 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |