missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse SF20/MYDGF Protein

Product Code. 18798846
Change view
Click to view available options
Quantità:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
18798846 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18798846 Supplier R&D Systems™ Supplier No. 10232MY050
 Articolo soggetto a restrizioni
Spedizione stimata: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Mouse SF20/MYDGF is immobilized at 2 μg/mL (100 μL/well), Recombinant Human Protein Disulfide Isomerase/P4HB binds with an ED50 of 0.7 to 4.2 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Numero di accesso Q9CPT4
Da utilizzare con (applicazione) Bioactivity
ID gene (immissione) 56005
Peso molecolare 16 kDa
Nome C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, Interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10
Quantità 50 μg
Sorgente Mouse myeloma cell line, NS0-derived mouse SF20/MYDGF protein Val25-Leu166
Requisiti di stoccaggio Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Simbolo del gene MYDGF
Coniugato Unconjugated
Specie Mouse
Ricostituzione Reconstitute at 500 μg/mL in PBS.
Tampone Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Livello di endotossine <0.10 EU per 1 μg of the protein by the LAL method.
Linea di prodotti R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Proteine SF20/MYDGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.