missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SIGIRR / TIR8 Protein
A DNA sequence encoding the Mus musculus (Mouse) SIGIRR / TIR8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8219-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_075546.2 | |
SIGIRR / TIR8 Protein | |
Research Use Only | |
Sigirr | |
Recombinant Protein | |
E. coli | |
MAGVCDMAPN FLSPSEDQAL GLALGREVAL NCTAWVFSRP QCPQPSVQWL KDGLALGNGS HFSLHEDFWV SANFSEIVSS VLVLNLTNAE DYGTFTCSVW NVSSHSFTLW RAGPAGH | |
Greater than 90% as determined by SDS-PAGE. |
24058 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |