missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TECTB Protein
A DNA sequence encoding the Mus musculus (Mouse) TECTB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9985-iv-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_033374.2 | |
TECTB Protein | |
Research Use Only | |
Tectb | |
Recombinant Protein | |
E. coli | |
KSCTPNKADV ILVFCYPKTI ITKIPECPYG WEVHQLALGG LCYNGVHEGG YYQFVIPDLS PKNKSYCGTQ SEYKPPIYHF YSHIVSNDST VIVKNQPVNY SFSCTYHSTY LVNQAAFDQR VATVHVKNGS MGTFESQLSL NFYTNAKFST KKEAPFVLET SEIGSDLFAG VEAKGLSVRF KVVLNSCWAT PSADFMYPLQ WQLINKGCPT DETVLVHENG KDHRATFQFN AFRFQNIPKL SKVWLHCETF ICDSEKLSCP VNCDKRKRML RDQTGGVLVV ELSLRSRA | |
Greater than 83.7% as determined by SDS-PAGE. |
21684 | |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |