missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Ubiquitin-protein ligase E3A Protein

A DNA sequence encoding the Mus musculus (Mouse) Ubiquitin-protein ligase E3A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6865-ec-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16056390

Quantità Prezzo
1 1245.00€ / 200µg
3 1160.96€ / 200µg
Spedizione stimata
21-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_035798.2
Ubiquitin-protein ligase E3A Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
22215
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Ube3a
Recombinant Protein
E. coli
HIIDDALVRL EMIAMENPAD LKKQLYVEFE GEQGVDEGGV SKEFFQLVVE EIFNPDIGMF TYDEATKLFW FNPSSFETEG QFTLIGILGL AIYNNCILDV HFPMVVYRKL MGKKGTFRDL GDSHPVLYQS LKDLLEYEGS VEDDMMITFQ ISQTDLFGNP MMYDLKENGD KIPITNENRK EFVISYSDYI LNKSVEKQFK AFRRGFHMVT NESPLKYLFR PEEIELLICG SRNLDFQALE ETTEYDGGYT RESVVIREFW EIVHSFTDEQ KRLFLLFTTG TDRAPVGGLG KLKMIIAKNG PDTERLPTSH TCFNVLLLPE YSSKEKLNV
Videos
Documenti