missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Oligoendopeptidase F homolog Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Oligoendopeptidase F homolog, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7236-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_109885.1 | |
Mycoplasma Oligoendopeptidase F homolog Protein | |
Research Use Only | |
pepF | |
Recombinant Protein | |
E. coli | |
MNNQYNWNLE VLLNGKSLAD NFTELKQLSE QEKALYDGGA CFQTKAKFTE FLQLQEKIEV LENRYSNFLS NKHAENSLDK TINDALFQYE MFKSEHALVF VDFEKNLFKH EKVIRAYLQD PALKQYQRDF ELVWRNKKHQ IDPASQKLLA QISPAWNQAD KIFNVLSTAD LNLQPVVYKG KTYVINAVSD YQSLLENKDR GLREAAYKVW | |
Greater than 90% as determined by SDS-PAGE. |
876899 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |