missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Bet v 2.0101 Protein
A cDNA sequence encoding the Bet v 2.0101 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11156-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Bet v 2.0101 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Bet v 2.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSWQTYVDEH LMCDIDGQAS NSLASAIVGH DGSVWAQSSS FPQFKPQEIT GIMKDFEEPG HLAPTGLHLG GIKYMVIQGE AGAVIRGKKG SGGITIKKTG QALVFGIYEE PVTPGQCNMV VERLGDYLID QGL |