missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other DnaK Lid Covering Subst Protein

A cDNA sequence encoding the DnaK Lid Covering Subst was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11685-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15948038

  • 1330.00€ / 1mg

Spedizione stimata: 31-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

DnaK Lid Covering Subst Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The DnaK protein contains 25mM Tris-HCl, pH 7.5, 100mM NaCl, 5mM DTT and 10%Glycerol.
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MNEDEIQKMV RDAEANAEAD RKFEELVQTR NQGDHLLHST RKQVEEAGDK LPADDKTAIESALTALETAL KGEDKAAIEA KMQELAQVSQ KLMEIAQQQH AQQQTAGADASANNAKDDDVVDAEFEEVKDKK
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali