missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Gly m 5.0101 Protein
A cDNA sequence encoding the Gly m 5.0101 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11998-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Gly m 5.0101 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VEKEECEEGE IPRPRPRPQH PEREPQQPGE KEEDEDEQPR PIPFPRPQPR QEEEHEQREE QEWPRKEEKR GEKGSEEEDE DEDEEQDERQ FPFPRPPHQK EERKQEEDED EEQQRESEES EDSELRRHKN KNPFLFGSNR FETLFKNQYG RIRVLQRFNQ RSPQLQNLRD YRILEFNSKP NTLLLPNHAD ADYLIVILNG TAILSLVNND DRDSYRLQSG DALRVPSGTT YYVVNPDNNE NLRLITLAIP VNKPGRFESF FLSSTEAQQS YLQGFSRNIL EASYDTKFEE INKVLFSREE GQQQGEQRLQ ESVIVEISKE QIRALSKRAK SSSRKTISSE DKPFNLRSRD PIYSNKLGKF FEITPEKNPQ LRDLDIFLSI VDMNEGALLL PHFNSKAIVI LVINEGDANI ELVGLKEQQQ EQQQEEQPLE VRKYRAELSE QDIFVIPAGY PVVVNATSNL NFFAIGINAE NNQRNFLAGS QDNVISQIPS QVQELAFPGS AQAVEKLLKN QRESYFVDAQ PKKKEEGNKG RKGPLSSILR AFY | |
Greater than 80.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Gly m 5.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |