missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other NEFM Protein
A cDNA sequence encoding the NEFM was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12825-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NEFM Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
MSYTLDSLGN PSAYRRVTET RSSFSRISGS PSSGFRSQSW SRGSPSTVSS SYKRSALAPR LTYSSAMLSS AESSLDFSQS SSLLDGGSGP GGDYKLSRSN EKEQIQGLND RFAGYIEKVH YLEQQNKEIE AEIQALRQKQ ASHAQLGDAY DQEIRELRAT LEMVNHEKAQ VQLDSDHLEE DIHRLKERFE EEARLRDDTE AAIRALRKDI EESSLVKVEL DKKVQSLQDE VAFLRSNHEE EVADLLAQIQ ASHITVERKD YLKTDISTAL KEIRSQLESH SDQNMHQAEE WFKCRYAKLT EAAEQNKEAI RSAKEEIAEY RRQLQSKSIE LESVRGTKES LERQLSDIEE RHNHDLSSYQ DTIQQLENEL RGTKWEMARH LREYQDLLNV KMALDIEIAA YRKLLEGEET RFSTFAGSIT GPLYTHRQPS IAISSKIQKT KVEAPKLKVQ HKFVEEIIEE TKVEDEKSEM EEALTAITEE LAVSVKEEVK EEEAEEKEEK EEAEEEVVAA KKSPVKATAP ELKEEEGEKE EEEGQEEEEE EEEAAKSDQA EEGGSEKEGS SEKEEGEQEE EGETEAEGEG EEAAAEAKEE KKMEEKAEEV APKEELAAEA KVEKPEKAKS PVAKSPTTKS PTAKSPEAKS PEAKSPTAKS PTAKSPVAKS PTAKSPEAKS PEAKSPTAKS PTAKSPAAKS PAPKSPVEEV KPKAEAGAEK GEQKEKVEEE KKEAKESPKE EKAEKKEEKP KDVPEKKKAE SPVKAESPVK EEVPAKPVKV SPEKEAKEEE KPQEKEKEKE KVEEVGGKEE GGLKESRKED IAINGEVEGK EEEQETKEKG SGGEEEKGVV TNGLDVSPGD EKKGGDKSEE KVVVTKMVEK ITSEGGDGAT KYITKSVTVT QKVEEHEETF EEKLVSTKKV | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The protein was lyophilized from a 1 mg/ml solution containing 10mM sodium phosphate, pH 7.5, 2mM DTT, 6M urea and 1mM EDTA. |