missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. Vaginalis P65 Protein
A cDNA sequence encoding the T. Vaginalis P65 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13679-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
T. Vaginalis P65 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
T. Vaginalis p65protein solution in PBS and 2M Urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Protein is >90% pure as determined by 12% PAGE (coomassie staining). |