missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. gingivalis Gingipain R2 Protein
A DNA sequence encoding the Porphyromonas gingivalis (strain ATCC BAA-308 / W83) Gingipain R2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6989-10ug
Additional Details : Peso : 0.01000kg
Specifications
WP_010956050.1 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
P. gingivalis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
P. gingivalis Gingipain R2 Protein | |
Research Use Only | |
rgpB | |
Recombinant Protein | |
E. coli | |
YTPVEEKENG RMIVIVPKKY EEDIEDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENII ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLANYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVAC | |
Greater than 90% as determined by SDS-PAGE. |