missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Epididymal secretory glutathione peroxidase Protein
A DNA sequence encoding the Sus scrofa (Pig) Epididymal secretory glutathione peroxidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6117-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_999051.1 | |
Epididymal secretory glutathione peroxidase Protein | |
Research Use Only | |
GPX5 | |
Recombinant Protein | |
Yeast | |
NSNLEKMDCY KDVTGTIYDY DAFTLNGNEH IQFKQYAGKH VLFVNVATYC GLTAQYPELN TLQEELKPFG LVVLGFPCNQ FGKQEPGENS EILLGLKYVR PGGGYVPNFQ LFEKGDVNGE KEQKVFTFLK HSCPHPSELI GSIGYISWEP IRVHDIRWNF EKFLVGPDGV PVMRWVHETP ISTVKSDILA YLKQFKTE | |
Greater than 90% as determined by SDS-PAGE. |
396920 | |
200 μg | |
Not determined. | |
Full Length of Mature Protein | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |