missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Uricase Protein
A DNA sequence encoding the Sus scrofa (Pig) Uricase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6871-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_999435.1 | |
Uricase Protein | |
Research Use Only | |
UOX | |
Recombinant Protein | |
E. coli | |
MAHYRNDYKK NDEVEFVRTG YGKDMIKVLH IQRDGKYHSI KEVATSVQLT LSSKKDYLHG DNSDVIPTDT IKNTVNVLAK FKGIKSIETF AVTICEHFLS SFKHVIRAQV YVEEVPWKRF EKNGVKHVHA FIYTPTGTHF CEVEQIRNGP PVIHSGIKDL KVLKTTQSGF EGFIKDQFTT LPEVKDRCFA TQVYCKWRYH QGRDVDFEAT WDTVRSIVLQ KFAGPYDKGE YSPSVQKTLY DIQVLTLGQV PEIEDMEISL PNIHYLNIDM SKMGLINKEE VLLPLDNPYG RITGTVKRKL TSRL | |
Greater than 90% as determined by SDS-PAGE. |
397510 | |
1 mg | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |