missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10637-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
GH Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAAGPRTSAL LAFALLCLPW TREVGAFPAM PLSSLFANAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKD EAQQRSDVEL LRFSLLLIQS WLGPVQFLSR VFTNSLVFGT SDRVYEKLKD LEEGIQALMR ELEDGSPRAG QILKQTYDKF DTNLRSDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFV ESSCAF |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Somatotropin contains 3 units/mg. | |
Porcine | |
Untagged | |
The Growth-hormone (1 mg/ml) was lyophilized after extensive dialyses against 0.34 mg sodium phosphate buffer (0.02 mg sodium phosphate monobasic and 0.32 mg sodium phosphate dibasic). |