missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Adipsin / Complement Factor D / CFD Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Adipsin / Complement Factor D / CFD, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5823-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001071110.1 | |
Adipsin / Complement Factor D / CFD Protein | |
Research Use Only | |
Cfd | |
Recombinant Protein | |
E. coli | |
ILGGQEAMAH ARPYMASVQV NGTHVCGGTL VDEQWVLSAA HCMDGVTKDE VVQVLLGAHS LSSPEPYKHL YDVQSVVLHP GSRPDSVEDD LMLFKLSHNA SLGPHVRPLP LQREDREVKP GTLCDVAGWG VVTHAGRRPD VLQQLTVSIM DRNTCNLRTY HDGAITKNMM CAESNRRDTC RGDSGGPLVC GDAVEAVVTW GSRVCGNRRK PGVFTRVATY VPWIENVLSG NVSVNVTA | |
Greater than 82.10% as determined by SDS-PAGE. |
54249 | |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |