missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Anionic trypsin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6536-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036767.1 | |
Anionic trypsin-1 Protein | |
Research Use Only | |
Prss1 | |
Recombinant Protein | |
E. coli | |
IVGGYTCPEH SVPYQVSLNS GYHFCGGSLI NDQWVVSAAH CYKSRIQVRL GEHNINVLEG DEQFINAAKI IKHPNYSSWT LNNDIMLIKL SSPVKLNARV APVALPSACA PAGTQCLISG WGNTLSNGVN NPDLLQCVDA PVLSQADCEA AYPGEITSSM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCALP DNPGVYTKVC NFVGWIQDTI AAN | |
Greater than 90% as determined by SDS-PAGE. |
24691 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |