missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GSTA1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) GSTA1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6122-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_058709.2 | |
GSTA1 Protein | |
Research Use Only | |
Gsta1 | |
Recombinant Protein | |
E. coli | |
SGKPVLHYFN ARGRMECIRW LLAAAGVEFD EKFIQSPEDL EKLKKDGNLM FDQVPMVEID GMKLAQTRAI LNYIATKYDL YGKDMKERAL IDMYTEGILD LTEMIMQLVI CPPDQKEAKT ALAKDRTKNR YLPAFEKVLK SHGQDYLVGN RLTRVDIHLL ELLLYVEEFD ASLLTSFPLL KAFKSRISSL PNVKKFLQPG SQRKLPVDAK QIEEARKIFK F | |
Greater than 90% as determined by SDS-PAGE. |
24422 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |